2-Methyldecahydroquinoline (mixture of isomers) ≥98.0%
Leverantör: TCI
|
Varning
|
M3189-1G
M3189-5G
TCIAM3189-1GEA
1675
SEK
TCIAM3189-1G
TCIAM3189-5G
2-Methyldecahydroquinoline (mixture of isomers) ≥98.0%
2-Methyldecahydroquinoline (mixture of isomers)
Formel:
C₁₀H₁₉N Kokpunkt: 206°C |
MDL Number:
MFCD20547663 CAS nummer: 20717-43-5 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % (total of isomer) |
Concentration (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...