2-Chloro-5-iodotoluene ≥98.0%
Leverantör: TCI
|
Varning
|
C3318-25G
C3318-5G
TCIAC3318-25GEA
4225
SEK
TCIAC3318-25G
TCIAC3318-5G
2-Chloro-5-iodotoluene ≥98.0%
2-Chloro-5-iodotoluene
Formel:
C₇H₆ClI Kokpunkt: 240°C Smältpunkt: 10°C |
MDL Number:
MFCD03093903 CAS nummer: 116632-41-8 |
Specifikation testresultat
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...