Chloropentafluoroacetone monohydrate ≥95.0%

Leverantör: TCI
Fara

C0993-1G C0993-5G
TCIAC0993-1GEA 685 SEK
TCIAC0993-1G TCIAC0993-5G
Chloropentafluoroacetone monohydrate ≥95.0%
Chloropentafluoroacetone monohydrate
Formel: C₃ClF₅O·H₂O
MDL Number: MFCD00150647
CAS nummer: 6984-99-2
UN: 2810
ADR: 6.1,III

Order Now

Specifikation testresultat

Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR