Dimetylsulfoxid, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Leverantör: Thermo Scientific
Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
348440010 348440025 348441000 348445000
348440010.EA 2275 SEK
348440010. 348440025. 348441000. 348445000.
Dimetylsulfoxid, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Dimetylsulfoxid
Formel: (CH₃)₂SO
MW: 78,14 g/mol
Kokpunkt: 189 °C (760 mmHg)
Smältpunkt: 18,4 °C
Densitet: 1,1 g/cm³
Flampunkt: 87 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002089
CAS nummer: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.70 %