Ethyl-3-hydroxyhexanoate ≥98.0%

Leverantör: TCI

Synonyms: Ethyl 3-hydroxyhexanoate

E0788-100G E0788-25G
TCIAE0788-100GEA 2400 SEK
TCIAE0788-100G TCIAE0788-25G
Ethyl-3-hydroxyhexanoate ≥98.0%
Ethyl-3-hydroxyhexanoate
Formel: C₈H₁₆O₃
MW: 160,21 g/mol
Kokpunkt: 92°C
Flampunkt: 94°C
MDL Number: MFCD00036604
CAS nummer: 2305-25-1
EINECS: 218-973-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9bcac6dba02f5985013211de18d491da362e9ad&vid=a9bcac6dba02f5985013211de18d491da362e9ad&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR